182192
Poly(lauryl methacrylate) solution
25 wt. % in toluene, analytical standard, average Mw 570,000 (Typical), average Mn 290,000 (Typical)
About This Item
Recommended Products
grade
analytical standard
Quality Level
mol wt
average Mn 290,000 (Typical)
average Mw 570,000 (Typical)
concentration
25 wt. % in toluene
technique(s)
gel permeation chromatography (GPC): suitable
format
neat
SMILES string
CCCCCCCCCCCCOC(=O)C(C)=C
General description
Other Notes
signalword
Danger
Hazard Classifications
Aquatic Chronic 3 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 Inhalation - STOT SE 3
target_organs
Central nervous system
Storage Class
3 - Flammable liquids
wgk_germany
WGK 3
flash_point_f
39.2 °F - closed cup
flash_point_c
4 °C - closed cup
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service