Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

181552

Supelco

Poly(isobutyl methacrylate)

analytical standard, for GPC, average Mw 300,000 (Typical), average Mn 140,000 (Typical)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C8H14O2)n
CAS Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard
for GPC

form

crystalline

mol wt

average Mn 140,000 (Typical)
average Mw 300,000 (Typical)

technique(s)

gel permeation chromatography (GPC): suitable

format

neat

SMILES string

CC(C)COC(=O)C(C)=C

InChI

1S/C8H14O2/c1-4-5-6-10-8(9)7(2)3/h2,4-6H2,1,3H3

InChI key

SOGAXMICEFXMKE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D Tantbirojn et al.
European journal of oral sciences, 108(6), 564-568 (2001-01-12)
An amphiphilic coating is configured as a substantive film that has a tendency for an in-plane two-dimensional polymerization. This coating is hypothesized to protect enamel from in vitro acid decalcification, assessed through the following artificial caries model. Three regions on
The effects of two temporary crown materials on the dental pulp of monkeys (Macaca fascicularis).
G J Pearson et al.
International endodontic journal, 19(3), 121-124 (1986-05-01)
Christopher C Godwin et al.
American journal of dentistry, 16(4), 260-266 (2003-10-29)
To provide a comprehensive assessment of the indoor environment of a representative dental office. Health and comfort parameters, including carbon dioxide (CO2), respirable particulate matter (PM2.5), volatile organic compounds (VOCs), temperature, relative humidity, light and sound level, were measured at
A G Zuccari et al.
Bio-medical materials and engineering, 7(5), 327-343 (1997-01-01)
Acrylic resins are commonly used in many dental applications; especially in the fabrication of provisional fixed partial dentures. Among noticed technical drawbacks associated with this material are unsatisfactory mechanical properties. Moreover, if acrylic resins are exposed to moist environment, water
Janaina Habib Jorge et al.
Quintessence international (Berlin, Germany : 1985), 40(10), e93-100 (2010-02-18)
To evaluate the effect of microwave postpolymerization heat treatment and water storage time on the cytotoxicity of denture base and reline acrylic resins. Sample disks of 6 acrylic resins were fabricated under aseptic conditions (Kooliner, GC; Tokuyama Rebase II, Tokuyama

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service