Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

18078

Supelco

4-Allylpyrocatechol

analytical standard

Synonym(s):

1,2-Dihydroxy-4-allylbenzene, 4-(2-Propenyl)-1,2-benzenediol, 4-Allyl-1,2-benzenediol, 4-Allylcatechol, Allylpyrocatechol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H10O2
CAS Number:
Molecular Weight:
150.17
Beilstein/REAXYS Number:
1937030
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

assay

≥95.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

Oc1ccc(CC=C)cc1O

InChI

1S/C9H10O2/c1-2-3-7-4-5-8(10)9(11)6-7/h2,4-6,10-11H,1,3H2

InChI key

FHEHIXJLCWUPCZ-UHFFFAOYSA-N

General description

4-Allylpyrocatechol also called Allylpyrocatechol is an antioxidant found in Piper Betel leaf extract. It is widely used as mouth freshener.[1] It is a phenolic compound, considered to be effective against indomethacin-gastropathy.[2]

application

It was used in a study performed to evaluate the phenolic content, antioxidant and antimicrobial activity of the bioactives of Piper Betle leaf extract using TLC-bioautography, HPTLC and GC-MS methods.[3]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

285.8 °F

flash_point_c

141 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Eisuke Kato et al.
Food chemistry, 136(3-4), 1239-1242 (2012-12-01)
Inhibition of pancreatic lipase is effective for a prevention of obesity. Eugenia polyantha is a tropical tree whose leaves are known as a spice and also as an ingredient for Jamu, the traditional medicine of Indonesia. We found inhibitory activity
M C Chang et al.
British journal of pharmacology, 152(1), 73-82 (2007-07-21)
Platelet hyperactivity is important in the pathogenesis of cardiovascular diseases. Betel leaf (PBL) is consumed by 200-600 million betel quid chewers in the world. Hydroxychavicol (HC), a betel leaf component, was tested for its antiplatelet effect. We tested the effect
C L Chen et al.
Cancer letters, 155(1), 29-35 (2000-05-18)
Hydroxychavicol (HC) is the major safrole urinary metabolite in rats and humans. The cytotoxic potential of HC in metabolically competent cells has yet to be studied. HC alone was slightly toxic to HepG2 cells. However, the cytotoxicity increased significantly (P<0.05)
TLC?bioautography-guided isolation, HPTLC and GC?MS-assisted analysis of bioactives of Piper betle leaf extract obtained from various extraction techniques: in vitro evaluation of phenolic content, antioxidant and antimicrobial activities.
Annegowda, H. V., et al.
Food Analytical Methods, 6.3, 715-726 (2013)
Anjali Pandey et al.
Cytokine, 49(1), 114-121 (2009-09-29)
The present study was undertaken to investigate the anti-arthritic activity of hydroxychavicol (HC) a major phenolic compound isolated from the aqueous extract leaves of plant Piper betle (Piperaceae). The compound showed significant lowering of pro-inflammatory (Th1) cytokine levels in arthritic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service