Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

18073

Supelco

2-Methylanisole

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4OCH3
CAS Number:
Molecular Weight:
122.16
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:

grade

analytical standard

assay

≥98.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.516 (lit.)
n20/D 1.518

bp

170-172 °C (lit.)

density

0.985 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

COc1ccccc1C

InChI

1S/C8H10O/c1-7-5-3-4-6-8(7)9-2/h3-6H,1-2H3

InChI key

DTFKRVXLBCAIOZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

It was used in a study done to understand peak distortion in RPLC due to viscosity differences between sample solvent and mobile phase.[1]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

pictograms

Flame

signalword

Warning

hcodes

Hazard Classifications

Flam. Liq. 3

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

125.6 °F - closed cup

flash_point_c

52 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Peak distortion in reversed-phase liquid chromatography as a consequence of viscosity differences between sample solvent and mobile phase.
Castells, Cecilia B., and Reynaldo C. Castells
Journal of Chromatography A, 805.1, 55-61 (1998)
Elodie Guyonnet Bilé et al.
ChemSusChem, 5(1), 91-101 (2012-01-18)
Optically active amphiphilic compounds derived from N-methylephedrine, N-methylprolinol, or cinchona derivatives possessing bromide or chiral lactate counterions were efficiently used as protective agents for rhodium(0) nanoparticles. The full characterization of these surfactants and the obtained nanocatalysts was performed by means
Graziela G Bianco et al.
The Journal of organic chemistry, 74(6), 2561-2566 (2009-02-24)
The first synthesis of the natural product (+)-mutisianthol was accomplished in 11 steps and in 21% overall yield from 2-methylanisole. The synthesis of its enantiomer was also performed in a similar overall yield. The absolute configuration of the sesquiterpene (+)-mutisianthol
L Higgins et al.
Archives of biochemistry and biophysics, 385(1), 220-230 (2001-05-22)
Regioselectivity is used to determine the absolute energetic differences for four different reactions catalyzed by P450. Abstraction of a hydrogen from a benzylic carbon containing a chlorine has a 1.0 kcal/mol lower barrier than abstraction from a simple benzylic carbon
Johannes Niebler et al.
Phytochemistry, 109, 66-75 (2014-12-04)
Frankincense has been known, traded and used throughout the ages for its exceptional aroma properties, and is still commonly used in both secular and religious settings to convey a pleasant odor. Surprisingly, the odoriferous principle(s) underlying its unique odor profile

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service