17403
2,3-Dihydroxy-biphenyl
derivatization grade (GC derivatization), LiChropur™, ≥98.0%
Synonym(s):
2,3-Biphenyldiol, 3-Phenylpyrocatechol
About This Item
grade
derivatization grade (GC derivatization)
Quality Level
assay
≥98.0% (HPLC)
≥98.0%
form
solid
quality
LiChropur™
reaction suitability
reagent type: derivatization reagent
reaction type: Alkylations
technique(s)
gas chromatography (GC): suitable
mp
108-113 °C
SMILES string
Oc1cccc(-c2ccccc2)c1O
InChI
1S/C12H10O2/c13-11-8-4-7-10(12(11)14)9-5-2-1-3-6-9/h1-8,13-14H
InChI key
YKOQAAJBYBTSBS-UHFFFAOYSA-N
General description
Legal Information
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service