Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

17293

Supelco

Lewatit® VP OC-1026

macroporous

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116105

form

macroporous

technique(s)

LPLC: suitable

loss

30-40% loss on drying

matrix

styrene-divinylbenzene

matrix active group

Carboxymethyl

particle size

0.3-1.25 mm

separation technique

cation exchange

Legal Information

Lewatit is a registered trademark of Lanxess Deutschland GmbH

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

One-Pot Biocatalytic Synthesis of Sugar Based Poly (ε-caprolactone)
Barrera-Rivera, Karla A., and Antonio Martinez-Richa
Macromolecular Symposia, 283, 144-151 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service