17281
1-(Pentafluoropropionyl)imidazole
derivatization grade (GC derivatization), ≥98.5%
Synonym(s):
PFPI, 1-(Perfluoropropionyl)imidazole
About This Item
Recommended Products
grade
derivatization grade (GC derivatization)
Quality Level
assay
≥98.5% (GC)
≥98.5%
reaction suitability
reagent type: derivatization reagent
reaction type: Esterifications
technique(s)
GC/MS: suitable
refractive index
n20/D 1.399
density
1.4 g/mL at 20 °C (lit.)
storage temp.
−20°C
SMILES string
FC(F)(F)C(F)(F)C(=O)n1ccnc1
InChI
1S/C6H3F5N2O/c7-5(8,6(9,10)11)4(14)13-2-1-12-3-13/h1-3H
InChI key
VZUSRIIEPFQBNE-UHFFFAOYSA-N
General description
Application
Other Notes
signalword
Danger
hcodes
Hazard Classifications
Acute Tox. 4 Oral - Skin Corr. 1B
Storage Class
8A - Combustible corrosive hazardous materials
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service