Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

17165

Millipore

MUG Tryptone Soya Agar

suitable for microbiology

Synonym(s):

CASO MUG Agar, Soybean Casein digest MUG Agar, Tryptone Soya MUG Agar

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171606

form

solid

Quality Level

shelf life

limited shelf life, expiry date on the label

composition

agar, 15 g/L
casein enzymic hydrolysate, 15 g/L
4-methylumbelliferyl-β-D-glucuronide (MUG), 0.1 g/L
papaic digest of soyabean meal, 5 g/L
sodium chloride, 5 g/L

final pH

7.3±0.2 (25 °C)

application(s)

cleaning products
clinical testing
cosmetics
environmental
food and beverages
food and beverages
food and beverages
personal care
veterinary

microbiology

suitability

nonselective and differential for Escherichia coli
nonselective and differential for bacteria (General Media)
nonselective and differential for coliforms

Application

For cultivation of fastidious and nonfastidious microorganisms by fluorogenic method.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service