17139
Ethyl Violet Azide Broth
suitable for microbiology, NutriSelect® Plus
Synonym(s):
EVA Broth
About This Item
Recommended Products
form
powder
Quality Level
shelf life
limited shelf life, expiry date on the label
composition
casein enzymic hydrolysate, 20 g/L
dextrose, 5 g/L
dipotassium phosphate, 2.7 g/L
ethyl violet, 0.00083 g/L
monopotassium phosphate, 2.7 g/L
sodiumazide, 0.4 g/L
sodium chloride, 5 g/L
manufacturer/tradename
NutriSelect® Plus
final pH
7.0±0.2 (25 °C)
application(s)
environmental
food and beverages
water monitoring
microbiology
suitability
selective for Enterococcus spp.
Application
Preparation Note
Footnote
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.
Legal Information
signalword
Warning
hcodes
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Aquatic Chronic 3
Storage Class
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Faceshields, Gloves, type P2 (EN 143) respirator cartridges
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service