About This Item
vapor density
<0.3 (25 °C, vs air)
vapor pressure
1 mmHg ( 146 °C)
description
Nominally 95-98% H2SO4
grade
puriss. p.a. plus
assay
≥95% (T)
ign. residue
≤0.0003%
bp
~290 °C (lit.)
density
1.840 g/mL at 25 °C (lit.)
anion traces
chloride (Cl-): ≤0.1 mg/kg
nitrate (NO3-): ≤0.1 mg/kg
phosphate (PO43-): ≤0.1 mg/kg
cation traces
Ag: ≤0.01 mg/kg
Al: ≤0.01 mg/kg
As, Sb (as As): ≤0.01 mg/kg
Au: ≤0.01 mg/kg
B: ≤0.01 mg/kg
Ba: ≤0.01 mg/kg
Be: ≤0.01 mg/kg
Bi: ≤0.01 mg/kg
Ca: ≤0.01 mg/kg
Cd: ≤0.01 mg/kg
Co: ≤0.01 mg/kg
Cr: ≤0.01 mg/kg
Cu: ≤0.01 mg/kg
Fe: ≤0.01 mg/kg
Ga: ≤0.01 mg/kg
Ge: ≤0.01 mg/kg
In: ≤0.01 mg/kg
K: ≤0.01 mg/kg
Li: ≤0.01 mg/kg
Mg: ≤0.01 mg/kg
Mn: ≤0.01 mg/kg
Mo: ≤0.01 mg/kg
NH4+: ≤0.5 mg/kg
Na: ≤0.01 mg/kg
Ni: ≤0.01 mg/kg
Pb: ≤0.01 mg/kg
Pt: ≤0.01 mg/kg
Sn: ≤0.01 mg/kg
Sr: ≤0.01 mg/kg
Ti: ≤0.01 mg/kg
Tl: ≤0.01 mg/kg
V: ≤0.01 mg/kg
Zn: ≤0.01 mg/kg
Zr: ≤0.01 mg/kg
SMILES string
OS(O)(=O)=O
InChI
1S/H2O4S/c1-5(2,3)4/h(H2,1,2,3,4)
InChI key
QAOWNCQODCNURD-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
signalword
Danger
hcodes
Hazard Classifications
Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A
Storage Class
8B - Non-combustible corrosive hazardous materials
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service