Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

16879

Supelco

Behenamide

analytical standard

Synonym(s):

Amide C22, Docosanamide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)20CONH2
CAS Number:
Molecular Weight:
339.60
Beilstein/REAXYS Number:
1792886
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

assay

≥98.0% (N)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

110-113 °C

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

functional group

amide

SMILES string

CCCCCCCCCCCCCCCCCCCCCC(N)=O

InChI

1S/C22H45NO/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22(23)24/h2-21H2,1H3,(H2,23,24)

InChI key

ORAWFNKFUWGRJG-UHFFFAOYSA-N

General description

Behenamides are primary fatty acid amides, which belong to fatty acyls subclass containing a carboxamide headgroup and an acyl tail with variable carbon length and unsaturation.[1]
This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food.

application

Behenamide may be used as a reference standard for the determination of behenamides in different biological samples by liquid chromatography coupled to tandem mass spectrometry (LC-MS/MS) in multiple reaction monitoring (MRM) mode.[1]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Determination of primary fatty acid amides in different biological fluids by LC-MS/MS in MRM mode with synthetic deuterated standards: influence of biofluid matrix on sample preparation.
Castillo-Peinado LS, et al.
Talanta, 29-36 (2018)
Tara Carpenter et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 809(1), 15-21 (2004-07-30)
Primary fatty acid amides (R-CO-NH2) and N-acylglycines (R-CO-NH-CH2-COOH) are classes of compounds that have only recently been isolated and characterized from biological sources. Key questions remain regarding how these lipid amides are produced and degraded in biological systems. Relative to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service