Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

16483

Sigma-Aldrich

β-tri-Calcium phosphate

puriss. p.a., ≥95% β-phase basis (sintered powder)

Synonym(s):

β-TCP, β-Tricalcium phosphate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Ca3O8P2
CAS Number:
Molecular Weight:
310.18
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
assay:
≥95% β-phase basis (sintered powder)
form:
powder

grade

puriss. p.a.

assay

≥95% β-phase basis (sintered powder)

form

powder

impurities

≤50 mg/kg total heavy metals (as lead)
≤500 mg/kg total sulfur (as SO4) (ICP)

anion traces

chloride (Cl-): ≤50 mg/kg

cation traces

As: ≤0.5 mg/kg
Ba: ≤20 mg/kg
Cd: ≤1 mg/kg
Co: ≤1 mg/kg
Cr: ≤15 mg/kg
Cu: ≤1 mg/kg
Hg: ≤0.5 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤20 mg/kg

InChI

1S/3Ca.2H3O4P/c;;;2*1-5(2,3)4/h;;;2*(H3,1,2,3,4)/q3*+2;;/p-6

Inchi Key

QORWJWZARLRLPR-UHFFFAOYSA-H

Looking for similar products? Visit Product Comparison Guide

Analysis Note

Phase Composition (by XRD analysis):
ß-TCP: >95% ; Hydroxylapatite: <2.0% ; a-TCP: <1.0%; TTCP: 0%
other Ca-P phases as Calcium pyrophosphate <2.0%

Other Notes

A -D suffix exists for administrative purposes only.
All -D packages are 100% the same product, same quality, same specification as the package sizes previously sold without a -D.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Wiltfang et al.
Journal of biomedical materials research, 63(2), 115-121 (2002-03-01)
In seven Goettingen minipigs 3.5--4.7-ml cancellous bone defects were created in the area of the tibial head on both sides. The defects were filled with alpha-TCP or beta-TCP (tricalciumphosphate). ITI implants (Straumann, Freiburg, Germany) of 3.2 x 12-mm length were
Development of calcium phosphate based bioceramics.
Sinha A, et al.
Bulletin of Materials Science, 24(6), 653-657 (2001)
M Muschik et al.
European spine journal : official publication of the European Spine Society, the European Spinal Deformity Society, and the European Section of the Cervical Spine Research Society, 10 Suppl 2, S178-S184 (2001-11-22)
The aim of this study is to evaluate the ability of beta-tricalcium phosphate (TCP) in granular form to achieve dorsal spondylodesis in adolescent idiopathic scoliosis (AIS). Twenty-eight patients underwent surgical correction and were followed up for 13+/-8 (range 6-33) months.
Evaluation of hydroxyapatite and β-tri calcium phosphate microplasma spray coated pin intra-medullary for bone repair in a rabbit model.
Dey A, et al.
Ceramics International, 37(4), 1377-1391 (2011)
Michael D Weiden et al.
PloS one, 9(10), e108476-e108476 (2014-10-02)
While exploring the effects of aerosol IFN-γ treatment in HIV-1/tuberculosis co-infected patients, we observed A to G mutations in HIV-1 envelope sequences derived from bronchoalveolar lavage (BAL) of aerosol IFN-γ-treated patients and induction of adenosine deaminase acting on RNA 1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service