Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

16460

Sigma-Aldrich

4-Bromobenzyl bromide

purum, ≥98.0% (GC)

Synonym(s):

α,4-Dibromotoluene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC6H4CH2Br
CAS Number:
Molecular Weight:
249.93
Beilstein/REAXYS Number:
606498
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

purum

assay

≥98.0% (GC)

form

solid

mp

60-63 °C
62-64 °C (lit.)

SMILES string

BrCc1ccc(Br)cc1

InChI

1S/C7H6Br2/c8-5-6-1-3-7(9)4-2-6/h1-4H,5H2

InChI key

YLRBJYMANQKEAW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

pictograms

Health hazardCorrosion

signalword

Danger

hcodes

Hazard Classifications

Resp. Sens. 1 - Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E V Sargent et al.
Drug and chemical toxicology, 22(4), 583-593 (1999-10-28)
As part of an occupational hazard evaluation, p-bromobenzyl bromide (p-BBB) was evaluated for genotoxic activity in the Ames microbial mutagenicity assay, the alkaline elution assay for DNA strand breaks in rat hepatocytes and the in vitro chromosome aberration assay in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service