Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

16252

Supelco

Poly(vinyl chloride) carboxylated

Selectophore, 5% carboxyl basis

Synonym(s):

Poly(vinyl chloride-co-acrylic acid)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C3H4O2 · C2H3Cl)x
CAS Number:
MDL number:
UNSPSC Code:
26111700
NACRES:
NB.61

Quality Level

description

for ion-selective electrodes

product line

Selectophore

mol wt

Mw ~165,000

concentration

5% (carboxyl)

SMILES string

ClC=C

InChI

1S/C3H4O2.C2H3Cl/c1-2-3(4)5;1-2-3/h2H,1H2,(H,4,5);2H,1H2

InChI key

SQNNHEYXAJPPKH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Carboxylated poly-vinyl chloride (PVC-COOH), is generally used in order to immobilize the potassium ion-selective and pH membrane electrodes. Carboxylated PVC-COOH membrane′s adhesive properties to an inert substrate such as polyimide-coated Kapton, can be greatly improved by reducing the plasticizer content of the membrane.[1]
Visit our Sensor Applications portal to learn more.

Application

Carboxylated PVC-COOH may be used as a polymeric matrix, for the immobilisation of the dopamine sensitive electrode, in an experimental study, done for the determination of dopamine by direct potentiometry in pharmaceutical preparations.[2]

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

José Pinela et al.
Food chemistry, 274, 686-690 (2018-10-31)
The suitability of post-packaging gamma radiation treatment for preserving total folates or vitamin B9 in watercress (Nasturtium officinale R. Br.) and buckler sorrel (Rumex induratus Boiss. & Reut.) during storage at 4 °C was evaluated. Comparable amounts of total folates were
Electroanalytical and biocompatibility studies on carboxylated poly (vinyl chloride) membranes for microfabricated array sensors.
Cosofret VV
Analyst, 119 (11), 2283-2292 (1994)
Dopamine ion-selective electrode for potentiometry in pharmaceutical preparations.
Lima JL
Microchimica Acta, 131 (3-4), 187-190 (1999)
IIIrd International Symposium on Neural Transplantation. Cambridge, 6-11 August, 1989. Proceedings.
Progress in brain research, 82, 1-734 (1990-01-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service