Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

16229

Sigma-Aldrich

1,5-Pentanediol

purum, ≥95.0% (GC)

Synonym(s):

Pentamethylene glycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO(CH2)5OH
CAS Number:
Molecular Weight:
104.15
Beilstein/REAXYS Number:
1560130
EC Number:
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:

vapor pressure

<0.01 mmHg ( 20 °C)

grade

purum

assay

≥95.0% (GC)

autoignition temp.

635 °F

expl. lim.

13.2 %

refractive index

n20/D 1.450 (lit.)
n20/D 1.450

bp

242 °C (lit.)

density

0.994 g/mL at 25 °C (lit.)

SMILES string

OCCCCCO

InChI

1S/C5H12O2/c6-4-2-1-3-5-7/h6-7H,1-5H2

InChI key

ALQSHHUCVQOPAS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

287.6 °F - closed cup

flash_point_c

142 °C - closed cup

ppe

Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Keisuke Yoshida et al.
Chemical communications (Cambridge, England), 48(55), 6981-6983 (2012-06-07)
Highly chemo- and regioselective acylation of 2-aminopentane-1,5-diol derivatives has been achieved by organocatalysis. An acyl group can be chemoselectively introduced onto the sterically hindered secondary hydroxy group in the presence of the primary one by virtue of the molecular recognition
Lai Wah Chan et al.
Biomaterials, 23(5), 1319-1326 (2002-01-26)
Calcium alginate microspheres were prepared by an emulsification method and cross-linked with various aldehydes using different methods. Methanal and pentanedial produced low aggregation of microspheres while octanal and octadecanal produced the opposite effect. The latter two aldehydes displaced very little
Jeanette Jacobsson Sundberg et al.
Expert opinion on investigational drugs, 17(4), 601-610 (2008-03-28)
The use of pentane-1,5-diol in topical pharmaceutical products is relatively new compared with, e.g., propane-1,2-diol (propylene glycol), also an aliphatic diol, which has been used for many years. Yet, what are the differences between diols in clinical efficacy, safety and
Treatment of atopic dermatitis with 1% hydrocortisone and 25% pentane-1,5-diol: effect on Staphylococcus aureus.
Mari Helen Sandström Falk et al.
Acta dermato-venereologica, 86(4), 372-373 (2006-07-29)
Keum Hwan Park et al.
Nanotechnology, 22(4), 045602-045602 (2010-12-16)
The size of silver nanocrystals in polyol synthesis can be simply controlled by tuning the viscosity of the reaction medium such as ethylene glycol, 1,2-propanediol, 1,4-butanediol and 1,5-pentanediol. We found that a higher viscose medium (1,5-pentanediol) led to monodispersed smaller

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service