Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

16180

Sigma-Aldrich

SODOSIL RA 12

alkaline, tensid-free cleaner

Sign Into View Organizational & Contract Pricing


About This Item

UBA Number:
08050039
UNSPSC Code:
12352000

form

solid

composition

phosphate, 15-30%

impurities

chloramine T
sodium hydroxide
sodium silicate

pH

12.5 ( in 0.3% caustic cleaning solution)

Legal Information

SODOSIL is a trademark of Sigma-Aldrich Chemie GmbH

signalword

Danger

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Resp. Sens. 1 - Skin Corr. 1 - STOT SE 3

target_organs

Respiratory system

supp_hazards

Storage Class

8B - Non-combustible corrosive hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service