Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

161403

Sigma-Aldrich

Mineral oil

suitable for IR spectroscopy

Synonym(s):

White mineral oil

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352100

form

viscous liquid

technique(s)

IR spectroscopy: suitable

refractive index

n20/D 1.467 (lit.)

density

0.838 g/mL at 25 °C (lit.)

InChI key

AEOVEGJBKQQFOP-DDVLFWKVSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Suitable for the preparation of nujol mulls in infrared analysis.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

No data available

flash_point_c

No data available

ppe

Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Keke Shao et al.
PloS one, 6(9), e24910-e24910 (2011-09-29)
Aptamers are short RNA or DNA oligonucleotides which can bind with different targets. Typically, they are selected from a large number of random DNA sequence libraries. The main strategy to obtain aptamers is systematic evolution of ligands by exponential enrichment
Takeshi Itabashi et al.
Nature methods, 6(2), 167-172 (2009-01-20)
Accurate chromosome segregation during meiosis depends on the assembly of a microtubule-based spindle of proper shape and size. Current models for spindle-size control focus on reaction diffusion-based chemical regulation and balance in activities of motor proteins. Although several molecular perturbations
Angela Rouyar et al.
PloS one, 6(10), e26443-e26443 (2011-10-27)
In nature the aerial trace of pheromone used by male moths to find a female appears as a train of discontinuous pulses separated by gaps among a complex odorant background constituted of plant volatiles. We investigated the effect of such
Xuchu Ma et al.
Dalton transactions (Cambridge, England : 2003), (46)(46), 10250-10255 (2009-11-19)
Na(4)Si(4) and Na(4)Ge(4) are ideal chemical precursors for inorganic clathrate structures, clusters, and nanocrystals. The monoclinic Zintl phases, Na(4)Si(4) and Na(4)Ge(4), contain isolated homo-tetrahedranide [Si(4)](4-) and [Ge(4)](4-) clusters surrounded by alkali metal cations. In this study, a simple scalable route
Eugene Y Kim et al.
Clinical immunology (Orlando, Fla.), 127(1), 98-106 (2008-02-16)
The pathogenesis of T cell-mediated diseases like rheumatoid arthritis (RA) has typically been explained in the context of the Th1-Th2 paradigm: the initiation/propagation by pro-inflammatory cytokines, and downregulation by Th2 cytokines. However, in our study based on the adjuvant-induced arthritis

Questions

1–3 of 3 Questions  
  1. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  2. What is Product 161403, Mineral oil, soluble in?

    1 answer
    1. This product is not soluble in water but is soluble in hydrocarbons. We typically do not perform solubility testing on this product.

      Helpful?

  3. What is the boiling point of Product 161403, Mineral oil?

    1 answer
    1. According to our supplier, the boiling point is not determined and we have not performed this testing.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service