Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

16100

Sigma-Aldrich

Bromoacetonitrile

purum, ≥97.0% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrCH2CN
CAS Number:
Molecular Weight:
119.95
Beilstein/REAXYS Number:
956569
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

purum

assay

≥97.0% (GC)

form

liquid

refractive index

n20/D 1.479 (lit.)
n20/D 1.479

bp

60-62 °C/24 mmHg (lit.)

density

1.722 g/mL at 25 °C (lit.)

SMILES string

BrCC#N

InChI

1S/C2H2BrN/c3-1-2-4/h1H2

InChI key

REXUYBKPWIPONM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Skull and crossbones

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

230.0 °F - closed cup

flash_point_c

110 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jongchan Kim et al.
American journal of cancer research, 8(9), 1823-1836 (2018-10-17)
β-catenin is not only a key component of adherens junctions but also a transcriptional co-activator downstream of canonical Wnt signaling. The Wnt/β-catenin pathway plays critical roles in animal development and tissue homeostasis, while mutation or overexpression of β-catenin often leads

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service