Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C17H20ClN3O · 0.5ZnCl2
CAS Number:
Molecular Weight:
385.96
UNSPSC Code:
12171500
EC Number:
279-675-0
Beilstein/REAXYS Number:
6035110
MDL number:
suitability
suitable for microscopy
SMILES string
Cl[Zn--](Cl)(Cl)Cl.CCN(CC)c1ccc2nc3cc(C)c(N)cc3[o+]c2c1.CCN(CC)c4ccc5nc6cc(C)c(N)cc6[o+]c5c4
Analysis Note
λmax. ∼622 nm, E(1%,1 cm) ∼1500-1700 (50% Ethanol)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.