Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

16007

Sigma-Aldrich

Brij® S10

main component: decaethylene glycol octadecyl ether

Synonym(s):

Polyethylene glycol octadecyl ether, Polyoxyethylene (10) stearyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C18H37(OCH2CH2)nOH, n~10
CAS Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

description

non-ionic

impurities

≤5% water

mp

37-39 °C (lit.)

HLB value

12.4

density

0.964 g/mL at 25 °C (lit.)

application(s)

cell analysis

InChI

1S/C20H41O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-19-22-20-18-21/h2-20H2,1H3

InChI key

PSEGVHKUPXNGGJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Brij is a registered trademark of Croda International PLC

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y C Lee et al.
Journal of pharmaceutical sciences, 86(4), 430-433 (1997-04-01)
An ocular insert is developed for the controlled systemic delivery of insulin. Commercially available Gelfoam absorbable gelatin sponge, USP, is used in the fabrication of the ocular insert in the form of a matrix system. Two eyedrop formulations and 13
N Albertson et al.
Journal of clinical microbiology, 36(5), 1232-1235 (1998-05-09)
The gastrointestinal pathogen Helicobacter pylori requires supplementation with either fetal calf serum (FCS), bovine serum albumin (BSA), or (2,6-dimethyl)-beta-cyclodextrin (CD) for growth in a complex or defined medium. Because the availability of medium in which all components were chemically defined
Anne Huhtala et al.
Journal of ocular pharmacology and therapeutics : the official journal of the Association for Ocular Pharmacology and Therapeutics, 19(1), 11-21 (2003-03-22)
This study was undertaken to investigate the use of the in vitro test WST-1, an assay of cell proliferation and viability, for a preliminary safety evaluation of topical ophthalmic preparations. The cytotoxicity of two surfactants, benzalkonium chloride (BAC) and polyoxyethylene-20-stearyl
Tatsuaki Tagami et al.
Journal of controlled release : official journal of the Controlled Release Society, 152(2), 303-309 (2011-02-23)
We have developed a novel and simplified thermosensitive liposomal formulation (HaT: Hyperthermia-activated cytoToxic) composed of DPPC lipid and Brij78 (96:4, molar ratio). The HaT nanoparticles were loaded with doxorubicin (DOX) with >95% efficiency when a pH gradient method and a
Jigna D Patel et al.
Pharmaceutical research, 24(2), 343-352 (2006-12-21)
The purpose of these studies was to prepare nanoparticles (NPs) with a small amount of surface-chelated nickel for obtaining enhanced binding of histidine-tagged (his-tag) proteins compared to non-histidine-tagged protein binding to charged nanoparticles. NPs were prepared from oil-in-water microemulsion precursors

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service