Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

15703

Supelco

Orange II-(naphthyl-d6) sodium salt

analytical standard

Synonym(s):

4-(2-Hydroxy-1-naphthyl-d6-azo)benzenesulfonic acid sodium salt, Acid Orange 7-d6, Acid Orange A-d6, Tropaeolin 000 No. 2-d6

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16D6H5N2NaO4S
CAS Number:
Molecular Weight:
356.36
UNSPSC Code:
85151701

grade

analytical standard

Quality Level

isotopic purity

≥95 atom % D (LC-MS)

assay

≥97.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

Storage temp.

2-8°C

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

STOT RE 1 Oral

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service