Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

154784

Sigma-Aldrich

1,2-Dichloroethane

spectrophotometric grade, ≥99%

Synonym(s):

Ethylene chloride, Ethylene dichloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCH2CH2Cl
CAS Number:
Molecular Weight:
98.96
Beilstein/REAXYS Number:
605264
EC Number:
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:

grade

spectrophotometric grade

vapor density

3.4 (20 °C, vs air)

vapor pressure

87 mmHg ( 25 °C)

assay

≥99%

form

liquid

autoignition temp.

775 °F

expl. lim.

15.6 %

technique(s)

UV/Vis spectroscopy: suitable

impurities

<0.020% water

evapn. residue

<0.002%

refractive index

n20/D 1.444 (lit.)

bp

83 °C (lit.)

mp

−35 °C (lit.)

density

1.256 g/mL at 25 °C (lit.)

λ

H2O reference

UV absorption

λ: 230 nm Amax: 1.00
λ: 240 nm Amax: 0.50
λ: 250 nm Amax: 0.15
λ: 300-400 nm Amax: 0.01

SMILES string

ClCCCl

InChI

1S/C2H4Cl2/c3-1-2-4/h1-2H2

InChI key

WSLDOOZREJYCGB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Asp. Tox. 1 - Carc. 1B - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

55.4 °F - closed cup

flash_point_c

13 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

François Perreault et al.
Chemical Society reviews, 44(16), 5861-5896 (2015-03-27)
Graphene-based materials are gaining heightened attention as novel materials for environmental applications. The unique physicochemical properties of graphene, notably its exceptionally high surface area, electron mobility, thermal conductivity, and mechanical strength, can lead to novel or improved technologies to address
Rongke Gao et al.
Biosensors & bioelectronics, 72, 230-236 (2015-05-20)
We report the application of a fully automated surface-enhanced Raman scattering (SERS)-based solenoid-embedded microfluidic device to the quantitative and sensitive detection of anthrax biomarker poly-γ-D-glutamic acid (PGA) in solution. Analysis is based on the competitive reaction between PGA and PGA-conjugated
Tom J Stockmann et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 17(47), 13206-13216 (2011-10-26)
The complexation reactions between strontium (Sr(2+)) and octyl(phenyl)-N,N-diisobutylcarbamoylmethylphosphine oxide (CMPO) were studied at the aqueous|1,2-dichloroethane (w|DCE) and aqueous|room-temperature ionic liquid (w|RTIL) microinterfaces, in order to understand its extraction in reprocessing spent nuclear fuels, remediation of environmental contamination, and potential radiological
Tatiana Wolfe et al.
Nanomedicine : nanotechnology, biology, and medicine, 11(5), 1277-1283 (2015-02-06)
We report potent radiosensitization of prostate cancers in vitro and in vivo using goserelin-conjugated gold nanorods. Progressive receptor-mediated internalization of conjugated nanorods over time increases the radiation interaction cross-section of cells and contributes to the effects observed in vitro. The
Peiyu Ge et al.
Proceedings of the National Academy of Sciences of the United States of America, 109(29), 11558-11563 (2012-06-06)
The photochemical reactivity of osmocene in a biphasic water-organic solvent system has been investigated to probe its water splitting properties. The photoreduction of aqueous protons to hydrogen under anaerobic conditions induced by osmocene dissolved in 1,2-dichloroethane and the subsequent water

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service