Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

154741

Sigma-Aldrich

Cyclohexane

ACS spectrophotometric grade, ≥99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12
CAS Number:
Molecular Weight:
84.16
Beilstein/REAXYS Number:
1900225
EC Number:
MDL number:
UNSPSC Code:
12191502
PubChem Substance ID:
grade:
ACS spectrophotometric grade
assay:
≥99%
technique(s):
UV/Vis spectroscopy: suitable
bp:
80.7 °C (lit.)
vapor pressure:
168.8 mmHg ( 37.7 °C)
77 mmHg ( 20 °C)

grade

ACS spectrophotometric grade

vapor density

2.9 (vs air)

vapor pressure

168.8 mmHg ( 37.7 °C)
77 mmHg ( 20 °C)

assay

≥99%

form

liquid

autoignition temp.

500 °F

expl. lim.

9 %

technique(s)

UV/Vis spectroscopy: suitable

impurities

H2SO4, passes test (darkened)
≤0.02% water

evapn. residue

<0.002%

color

APHA: ≤10

refractive index

n20/D 1.426 (lit.)

bp

80.7 °C (lit.)

mp

4-7 °C (lit.)

density

0.779 g/mL at 25 °C (lit.)

λ

H2O reference

UV absorption

λ: 210 nm Amax: 1.00
λ: 220 nm Amax: 0.50
λ: 230 nm Amax: 0.20
λ: 240 nm Amax: 0.08
λ: 250 nm Amax: 0.03
λ: 260 nm Amax: 0.02
λ: 300-400 nm Amax: 0.01

SMILES string

C1CCCCC1

InChI

1S/C6H12/c1-2-4-6-5-3-1/h1-6H2

InChI key

XDTMQSROBMDMFD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

signalword

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Central nervous system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

-4.0 °F - closed cup

flash_point_c

-20 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Omid D Rahmanian et al.
Microfluidics and nanofluidics, 18(5-6), 1045-1053 (2015-05-15)
A simple and reliable method for fabricating single-use normally closed burst valves in thermoplastic microfluidic devices is presented, using a process flow that is readily integrated into established workflows for the fabrication of thermoplastic microfluidics. An experimental study of valve
Lei Zhou et al.
Nature communications, 6, 6938-6938 (2015-04-25)
The identification of potential diagnostic markers and target molecules among the plethora of tumour oncoproteins for cancer diagnosis requires facile technology that is capable of quantitatively analysing multiple biomarkers in tumour cells and tissues. Diagnostic and prognostic classifications of human
Amanda L Pitts et al.
Journal of the American Chemical Society, 136(24), 8614-8625 (2014-05-16)
Carbon-hydrogen bond activation reactions of four cycloalkanes (C5H10, C6H12, C7H14, and C8H16) by the Cp'Rh(CO) fragments (Cp' = η(5)-C5H5 (Cp) or η(5)-C5Me5 (Cp*)) were modeled theoretically by combining density functional and coupled cluster theories, and their reaction rates were measured
Robert Duran et al.
Environmental science and pollution research international, 22(20), 15215-15229 (2015-02-26)
The exploration of marine Actinobacteria has as major challenge to answer basic questions of microbial ecology that, in turn, will provide useful information to exploit Actinobacteria metabolisms in biotechnological processes. The ecological functions performed by Actinobacteria in marine sediments are
Rajiv L Gaur et al.
PLoS pathogens, 10(9), e1004376-e1004376 (2014-09-19)
Mycobacterium tuberculosis employs various virulence strategies to subvert host immune responses in order to persist and cause disease. Interaction of M. tuberculosis with mannose receptor on macrophages via surface-exposed lipoarabinomannan (LAM) is believed to be critical for cell entry, inhibition

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service