154679
1-Butanol
spectrophotometric grade, 99.5%
Synonym(s):
n-Butanol, Butyl alcohol
About This Item
Recommended Products
grade
spectrophotometric grade
vapor density
2.55 (vs air)
vapor pressure
5 (4 mmHg) at 20 °C hPa
assay
99.5%
form
liquid
autoignition temp.
649 °F
expl. lim.
11.2 %
technique(s)
UV/Vis spectroscopy: suitable
impurities
≤0.1% water (Karl Fischer)
evapn. residue
≤0.0005%
color
APHA: ≤10
refractive index
n20/D 1.399 (lit.)
bp
116-118 °C (lit.)
mp
−90 °C (lit.)
density
0.81 g/mL at 25 °C (lit.)
λ
H2O reference
UV absorption
λ: 220 nm Amax: 1.0
λ: 225 nm Amax: 0.50
λ: 240 nm Amax: 0.10
λ: 260 nm Amax: 0.04
λ: 280-400 nm Amax: 0.01
SMILES string
CCCCO
InChI
1S/C4H10O/c1-2-3-4-5/h5H,2-4H2,1H3
InChI key
LRHPLDYGYMQRHN-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Other Notes
All -D packages are 100% the same product, same quality, same specification as the package sizes previously sold without a -D.
signalword
Danger
Hazard Classifications
Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3
target_organs
Central nervous system, Respiratory system
Storage Class
3 - Flammable liquids
wgk_germany
WGK 1
flash_point_f
95.0 °F - Pensky-Martens closed cup
flash_point_c
35 °C - Pensky-Martens closed cup
ppe
Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service