15358
Blotting-Nylon 66 membranes, type B, positive
roll
Synonym(s):
Nylon 66-Blotting membranes, type B
About This Item
Recommended Products
grade
Molecular Biology
form
roll
membrane size
0.3 × 3.3 m , rolled up
surface coverage
surface (quaternary ammonium groups) ≥99%
pore size
0.45 μm pore size
storage temp.
room temp
Looking for similar products? Visit Product Comparison Guide
General description
Application
Other Notes
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service