Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

15337

Supelco

Lead ionophore III

Selectophore

Synonym(s):

ETH 5435, N,N,N′,N′-Tetradodecyl-3,6-dioxaoctanedithioamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C54H108N2O2S2
CAS Number:
Molecular Weight:
881.58
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:

product line

Selectophore

SMILES string

CCCCCCCCCCCCN(CCCCCCCCCCCC)C(=S)COCCOCC(=S)N(CCCCCCCCCCCC)CCCCCCCCCCCC

InChI

1S/C54H108N2O2S2/c1-5-9-13-17-21-25-29-33-37-41-45-55(46-42-38-34-30-26-22-18-14-10-6-2)53(59)51-57-49-50-58-52-54(60)56(47-43-39-35-31-27-23-19-15-11-7-3)48-44-40-36-32-28-24-20-16-12-8-4/h5-52H2,1-4H3

InChI key

ZUONOYYCMQYKBW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Visit our Sensor Applications portal to learn more.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Physical form

solution in 0.5 mL heptane

Other Notes

Lipophilic lead ionophore employed in optode membranes with subnanomolar detection limits[1]

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

Flame

signalword

Danger

hcodes

pcodes

Hazard Classifications

Flam. Liq. 2

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

24.8 °F - closed cup

flash_point_c

-4 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T. Sokalski et al.
Journal of the American Chemical Society, 119, 11347-11347 (1997)
M. Lerchi et al.
Analytical Chemistry, 64, 1534-1534 (1992)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service