Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

14673

Supelco

7-Methylcoumarin

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H8O2
CAS Number:
Molecular Weight:
160.17
EC Number:
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

analytical standard

Quality Level

assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

bp

171-172 °C/11 mmHg (lit.)

mp

128-130 °C (lit.)

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

SMILES string

Cc1ccc2C=CC(=O)Oc2c1

InChI

1S/C10H8O2/c1-7-2-3-8-4-5-10(11)12-9(8)6-7/h2-6H,1H3

InChI key

DLHXRDUXNVEIEY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

7-Methylcoumarin is a substituted coumarin, which belongs to the class of heterocyclic oxygen compounds with benzopyran-2-one moiety as the core. It is commonly present as a component of food and beverage products.

Application

7-Methylcoumarin may be used as an analytical standard for the determination of the analyte in cosmetics, tobacco products, and plant extracts by liquid chromatography (LC) based techniques.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simultaneous determination of 17 coumarins in cosmetics by high performance liquid chromatography
Xi HW, et al.
Journal of Instrumental Analysis, 12 (2010)
Zhiqin Ren et al.
Molecules (Basel, Switzerland), 21(11) (2016-11-12)
In this paper an analytical method based on high performance liquid chromatography coupled to tandem mass spectrometry (HPLC-MS/MS) for the determination of coumarin and its derivatives in tobacco products was developed. The MS/MS fragmentation pathways of the eight coumarins were
A study of the analytical behaviour of selected synthetic and naturally occurring coumarins using liquid chromatography, ion trap mass spectrometry, gas chromatography and polarography and the construction of an appropriate database for coumarin character
Smyth WF, et al.
Analytica Chimica Acta, 564(2), 201-210 (2006)
A Cappiello et al.
Journal of the American Society for Mass Spectrometry, 6(2), 132-139 (1995-02-01)
Coumarins are a large group of compounds that are naturally present in plant tissues and that exhibit a wide range of pharmacological properties. Analytical methods based on chromatographic techniques and conventional detectors are inadequate to accurately analyze coumarins in complex
Determination of Coumarins in Cosmetics with High Performance Liquid Chromatography
Zhao XY, et al.
Journal of analytical science, 27, 49-52 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service