Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

14643

Supelco

16-Hexadecanolide

analytical standard

Synonym(s):

Juniperic acid lactone, NSC 33546

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H30O2
CAS Number:
Molecular Weight:
254.41
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

assay

≥98.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

34-38 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

O=C1CCCCCCCCCCCCCCCO1

InChI

1S/C16H30O2/c17-16-14-12-10-8-6-4-2-1-3-5-7-9-11-13-15-18-16/h1-15H2

InChI key

LOKPJYNMYCVCRM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fei-Meng Zheng et al.
BMB reports, 52(9), 566-571 (2019-08-14)
Lymphoma is one of the most curable types of cancer. However, drug resistance is the main challenge faced in lymphoma treatment. Peroxisomal acyl-CoA oxidase 1 (ACOX1) is the rate-limiting enzyme in fatty acid β-oxidation. Deregulation of ACOX1 has been linked
Jianxia Wen et al.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie, 115, 108881-108881 (2019-04-28)
Higenamine (HG) is a well-known selective activator of beta2-adrenergic receptor (β2-AR) with a positive inotropic effect. The present study showed that HG combined with [6]-gingerol (HG/[6]-GR) protects H9c2 cells from doxorubicin (DOX)-induced mitochondrial energy metabolism disorder and respiratory dysfunction. H9c2
Cinsley Gentillon et al.
Journal of molecular and cellular cardiology, 132, 120-135 (2019-05-15)
Immature phenotypes of cardiomyocytes derived from human induced pluripotent stem cells (hiPSC-CMs) limit the utility of these cells in clinical application and basic research. During cardiac development, postnatal cardiomyocytes experience high oxygen tension along with a concomitant downregulation of hypoxia-inducible
Heli Routti et al.
Scientific reports, 9(1), 6918-6918 (2019-05-08)
Peroxisome proliferator-activated receptor alfa (PPARA/NR1C1) is a ligand activated nuclear receptor that is a key regulator of lipid metabolism in tissues with high fatty acid catabolism such as the liver. Here, we cloned PPARA from polar bear liver tissue and
Yujia Han et al.
Biomolecules & therapeutics, 28(5), 397-404 (2020-06-25)
Adiponectin secretion-promoting compounds have therapeutic potentials in human metabolic diseases. Diallyl biphenyl-type neolignan compounds, magnolol, honokiol, and 4-O-methylhonokiol, from a Magnolia officinalis extract were screened as adiponectin- secretion promoting compounds in the adipogenic differentiation model of human bone marrow mesenchymal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service