Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

14465

Supelco

Proquinazid-(propoxy-d7)

PESTANAL®, analytical standard

Synonym(s):

6-Iodo-2-propoxy-d7-3-propyl-4(3H)-quinazolinone, Proquinazid-d7)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14D7H10IN2O2
Molecular Weight:
379.24
UNSPSC Code:
12352200
PubChem Substance ID:

grade

analytical standard

Quality Level

product line

PESTANAL®

assay

≥98.0% (GC)

shelf life

limited shelf life, expiry date on the label

application(s)

agriculture

format

neat

storage temp.

2-8°C

SMILES string

O=C1C2=CC(I)=CC=C2N=C(OC([2H])([2H])C([2H])([2H])C([2H])([2H])[2H])N1CCC

General description

Proquinazid-(propoxy-d7) is an isotope-labeled analog of the quinazolinone fungicide proquinazid, wherein the propoxy protons are replaced by deuterium.

Application

Isotope-labeled compounds are increasingly used in isotope dilution mass spectrometry (IDMS) for the quantitative analysis of pesticides. The main advantage of using the IDMS technique is because isotope-labeled compounds have nearly the same physical properties as their non-labeled counterpart analog, and thus show identical behavior during the workup and sample preparation process. This helps in overcoming the problems of matrix effects generally encountered in the usual LC-MS/GC-MS analysis potentially resulting in biased results. By spiking the sample before workup with its isotope labeled analog, the loss of analyte that leads to matrix effects can be determined and compensated.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

pictograms

Health hazardEnvironment

signalword

Warning

hcodes

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Baseline sensitivity to proquinazid in Blumeria graminis f. sp. tritici and Erysiphe necator and cross-resistance with other fungicides
Genet JL and Jaworska G
Pest Management Science, 65(8), 878-884 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service