Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

14405

Sigma-Aldrich

(+)-Biotin N-hydroxysuccinimide ester

≥95% (HPLC)

Synonym(s):

(+)-Biotin N-succinimidyl ester, d-Biotin NHS ester, BNHS, Biotinyl-N-hydroxy-succinimide, N-Hydroxysuccinimidobiotin, N-Succinimidyl D-biotinate, NHS-Biotin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H19N3O5S
CAS Number:
Molecular Weight:
341.38
Beilstein/REAXYS Number:
4720781
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

biological source

synthetic

Quality Level

assay

≥95% (HPLC)

form

powder

optical activity

[α]20/D +53±1°, c = 1% in DMF

color

white to off-white

mp

212-214 °C

storage temp.

−20°C

SMILES string

[H][C@]12CS[C@@H](CCCCC(=O)ON3C(=O)CCC3=O)[C@@]1([H])NC(=O)N2

InChI

1S/C14H19N3O5S/c18-10-5-6-11(19)17(10)22-12(20)4-2-1-3-9-13-8(7-23-9)15-14(21)16-13/h8-9,13H,1-7H2,(H2,15,16,21)/t8-,9-,13-/m0/s1

InChI key

YMXHPSHLTSZXKH-RVBZMBCESA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Activated biotin, generally used for attaching biotin to amino acids, peptides and proteins[1]; Review: The use of the avidin-biotin complex as a tool in molecular biology[2]; On-resin biotinylation of chemically synthesized proteins for one-step purification[3]

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maria Livia Bernardi et al.
PloS one, 6(11), e27856-e27856 (2011-11-25)
Food allergy is increasingly common worldwide. Tools for allergy diagnosis measuring IgE improved much since allergenic molecules and microarrays started to be used. IgE response toward allergens belonging to the same group of molecules has not been comprehensively explored using
Yasuko Hirakawa et al.
Journal of molecular signaling, 4, 6-6 (2009-09-04)
Glucocorticoid hormones (GCs) induce apoptosis of leukemic T-cells by transcriptional regulation via the GC receptor, GR. In the human leukemic CEM cell culture model, RCAN1 has been identified as one of the genes that is specifically upregulated only in the
Xiao-Lin Luo et al.
Cancer research, 78(13), 3532-3543 (2018-05-08)
A high rate of disease relapse makes epithelial ovarian cancer (EOC) the leading cause of death among all gynecologic malignancies. These relapses are often due to tumor dormancy. Here we identify the RNA polymerase II transcriptional mediator subunit 12 (MED12)
Ran Li et al.
Molecular cancer therapeutics, 17(1), 169-182 (2017-09-25)
In response to cellular stress, phosphatidylserine is exposed on the outer membrane leaflet of tumor blood vessels and cancer cells, motivating the development of phosphatidylserine-specific therapies. The generation of drug-conjugated phosphatidylserine-targeting agents represents an unexplored therapeutic approach, for which antitumor
Tue W Kragstrup et al.
Arthritis research & therapy, 16(1), R42-R42 (2014-02-05)
Spondyloarthritis (SpA) comprises a group of diseases often associated with HLA-B27 and characterized by inflammation of the entheses and joints of the axial skeleton. The inflammatory process in SpA is presumably driven by innate immune cells but is still poorly

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service