Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

13735

Sigma-Aldrich

(R)-(+)-3-Benzyloxy-1,2-propanediol

puriss., ≥99.0% (sum of enantiomers, GC)

Synonym(s):

(R)-Glycerol 1-benzyl ether, 3-Benzyl-sn-glycerol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H14O3
CAS Number:
Molecular Weight:
182.22
Beilstein/REAXYS Number:
2048922
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

grade

puriss.

assay

≥99.0% (sum of enantiomers, GC)

form

solid

optical activity

[α]20/D +5.5±1.0°, c = 20% in chloroform

storage temp.

2-8°C

SMILES string

OC[C@@H](O)COCc1ccccc1

InChI

1S/C10H14O3/c11-6-10(12)8-13-7-9-4-2-1-3-5-9/h1-5,10-12H,6-8H2/t10-/m1/s1

InChI key

LWCIBYRXSHRIAP-SNVBAGLBSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Important starting material for the synthesis of lipids, e.g. by diacylation or dialkylation [1]; 1-mono-derivatization [2]; Selective protection of primary OH [3]

replaced by

Product No.
Description
Pricing

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K.C. Schneider et al.
Journal of the American Chemical Society, 112, 453-453 (1990)
Synthesis of labeled phospholipids in high yield.
H Eibl et al.
Methods in enzymology, 98, 623-632 (1983-01-01)
G. Hirth et al.
Helvetica Chimica Acta, 65, 1059-1059 (1982)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service