13593
(−)-N-Benzyl-N-methylephedrinium bromide
purum, ≥99.0% (AT)
Synonym(s):
Benzyl(2-hydroxy-1-methyl-2-phenylethyl)dimethylammonium bromide
About This Item
grade
purum
assay
≥99.0% (AT)
form
solid
optical activity
[α]20/D −5.3±0.5°, c = 4.5% in methanol
mp
209-211 °C (lit.)
209-211 °C
SMILES string
[Br-].C[C@@H]([C@H](O)c1ccccc1)[N+](C)(C)Cc2ccccc2
InChI
1S/C18H24NO.BrH/c1-15(18(20)17-12-8-5-9-13-17)19(2,3)14-16-10-6-4-7-11-16;/h4-13,15,18,20H,14H2,1-3H3;1H/q+1;/p-1/t15-,18-;/m0./s1
InChI key
DSUGMMVBRSBBCH-NKGQWRHHSA-M
Looking for similar products? Visit Product Comparison Guide
Other Notes
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service