Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

13453

Sigma-Aldrich

3-Benzyl-5-(2-hydroxyethyl)-4-methylthiazolium chloride

purum, ≥99.0% (AT)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H16ClNOS
CAS Number:
Molecular Weight:
269.79
Beilstein/REAXYS Number:
3788921
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

grade

purum

assay

≥99.0% (AT)

form

solid

mp

142-144 °C (lit.)

SMILES string

[Cl-].Cc1c(CCO)sc[n+]1Cc2ccccc2

InChI

1S/C13H16NOS.ClH/c1-11-13(7-8-15)16-10-14(11)9-12-5-3-2-4-6-12;/h2-6,10,15H,7-9H2,1H3;1H/q+1;/p-1

InChI key

IWSVLBKHBJGMAA-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Other Notes

Catalyst for the addition of aliphatic and heterocyclic aldehydes to α,β-unsaturated ketones, nitriles and esters[1][2][3]

replaced by

Product No.
Description
Pricing

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H. Stetter et al.
Organic Syntheses, 62, 170-170 (1984)
H. Stetter et al.
Angewandte Chemie (International Edition in English), 88, 695-695 (1976)
H. Stetter et al.
Org. React., 40, 407-407 (1991)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service