Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

134270

AOCS-Tintometer Colour Reference Standard

3.0R 22Y (5¼″)

Synonym(s):

AOCS-Tintometer Color Reference Standard

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

analytical standard

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Paragon Scientific Ltd

application(s)

cleaning products
coatings
cosmetics
environmental
flavors and fragrances
food and beverages
glass & ceramic
personal care
pharmaceutical

format

mixture

Looking for similar products? Visit Product Comparison Guide

General description

AOCS-Tintometer colour reference standard is an analytical standard for colour solutions, manufactured and tested according to ISO 9001 Quality management system.

Please note that the value stated on this page is nominal. All certified values for the current lots of all Tintometer colour reference standards (AOCS) can be found on the Paragon Scientific webpage.

Application

Used to calibrate, control and qualify colour measurement instruments such as colorimeters.

Features and Benefits

  • Available with a certificate of calibration and safety data sheet
  • Supplied in high-quality 500-mL tamper-evident packaging
  • Full traceability to international standards with maximum data accuracy
  • 12-months shelf life

pictograms

Health hazardCorrosion

signalword

Danger

Hazard Classifications

Aquatic Chronic 3 - Carc. 1B - Met. Corr. 1

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service