Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

134250

AOCS-Tintometer Colour Reference Standard

1.3R 8.8Y (5¼″)

Synonym(s):

AOCS-Tintometer Color Reference Standard

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

analytical standard

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Paragon Scientific Ltd

bp

100 °C

application(s)

cleaning products
coatings
cosmetics
environmental
flavors and fragrances
food and beverages
glass & ceramic
personal care
pharmaceutical

format

mixture

Looking for similar products? Visit Product Comparison Guide

General description

AOCS-Tintometer colour reference standard is ideal for routine monitoring of colour measuring instruments and validation of test data. This product is certified under ISO 9001 Quality system.

Please note that the value stated on this page is nominal. All certified values for the current lots of all Tintometer colour reference standards (AOCS) can be found on the Paragon Scientific webpage.

Application

This product is used in the regular checking of colorimeters used to analyze colour control of oils, paint vehicles, chemicals, coatings, etc.

Features and Benefits

  • Accompanied with a certificate of calibration, conforming to ISO 9001 quality management system
  • Tamper-evident security packaging, assuring sample integrity
  • Full traceability to international standards with maximum data accuracy
  • Supplied in a 500 mL bottle with a 12-month guarantee of colour stability and full documentation including MSDS

pictograms

Health hazardCorrosion

signalword

Danger

Hazard Classifications

Aquatic Chronic 3 - Carc. 1B - Met. Corr. 1

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service