Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

134220

Gardner Colour Reference Standard

ASTM D 1544, D 6166 (ASTM colour 8)

Synonym(s):

Color Reference Standard Gardner 8, Colour Reference Standard Gardner 8, Gardner Color Reference Standard

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

agency

according to ASTM® D 1544
according to ASTM® D 6166

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Paragon Scientific Ltd

application(s)

cleaning products
coatings
cosmetics
environmental
flavors and fragrances
food and beverages
glass & ceramic
personal care
pharmaceutical

format

mixture

Looking for similar products? Visit Product Comparison Guide

General description

This product is a certified reference material (CRM) certified to international standards BS EN ISO / IEC 17025 and ISO 17034 under UKAS accreditation.

Please note that the value stated on this page is nominal. All certified values for the current lots of all Gardner colour reference standards can be found on the Paragon Scientific webpage.

Application

CRM tested according to ASTM D 1544, D 6166 methods is suitable for use in calibration of colour measuring apparatus and verification of test results. Its main application is in process industries that employ colorimeters to study the colour control of products such as edible and industrial oils, fuels, chemicals, pharmaceuticals, paints and coatings.

Features and Benefits

  • Produced and tested in accordance with ISO 17025 / ISO 17034 under UKAS accreditation
  • Full Traceability to international standards
  • Colour values in ASTM D 1544/D 6166 colour range
  • Supplied in a 500-mL tamper-evident bottle
  • 18-months shelf life
  • Available with CoA and MSDS

Legal Information

ASTM is a registered trademark of American Society for Testing and Materials

replaced by

pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Asp. Tox. 1

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kim A Ngo et al.
Cell reports, 30(8), 2758-2775 (2020-02-27)
Nuclear factor κB (NF-κB) RelA is the potent transcriptional activator of inflammatory response genes. We stringently defined a list of direct RelA target genes by integrating physical (chromatin immunoprecipitation sequencing [ChIP-seq]) and functional (RNA sequencing [RNA-seq] in knockouts) datasets. We
Eugen Dhimolea et al.
Cancer cell, 39(2), 240-256 (2021-01-09)
Treatment-persistent residual tumors impede curative cancer therapy. To understand this cancer cell state we generated models of treatment persistence that simulate the residual tumors. We observe that treatment-persistent tumor cells in organoids, xenografts, and cancer patients adopt a distinct and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service