Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

134050

Saybolt Colour Reference Standard

ASTM D 6045, D 156 (ASTM colour 0)

Synonym(s):

Colour Reference Standard Saybolt 0, Saybolt Color Reference Standard

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

agency

according to ASTM® D 156
according to ASTM® D 6045

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Paragon Scientific Ltd

application(s)

cleaning products
coatings
cosmetics
environmental
flavors and fragrances
food and beverages
glass & ceramic
personal care
pharmaceutical

format

mixture

Looking for similar products? Visit Product Comparison Guide

General description

Saybolt colour (ASTM colour 0) reference standard is a certified reference material (CRM), traceable to international standards BS EN ISO / IEC 17025 and ISO 17034 under UKAS accreditation.

Please note that the value stated on this page is nominal. All certified values for the current lots of all Saybolt colour reference standards can be found on the Paragon Scientific webpage.

Application

Colour Reference Standard Saybolt 0 is used to standardize colour measuring equipment and validate test results.

It may be used as a colour standard in spectroscopic methods to analyze the quality of petroleum products.

Features and Benefits

  • Full Traceability to international standards with maximum data accuracy
  • Colour values compliant to ASTM D 6045 and D 156 colour ranges
  • 500-mL pack size available with safety data sheet
  • 18-months shelf life
  • Tamper-evident security packaging

Legal Information

ASTM is a registered trademark of American Society for Testing and Materials

replaced by

pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Asp. Tox. 1 - Carc. 1B

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service