Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

134020

ASTM Colour Reference Standard

ASTM D 6045, D1500 (ASTM colour 5)

Synonym(s):

ASTM Color Reference Standard, Color Reference Standard ASTM Color 5, Colour Reference Standard ASTM Colour 5

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

agency

according to ASTM® D 6045
according to ASTM® D1500

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Paragon Scientific Ltd

application(s)

cleaning products
coatings
cosmetics
environmental
flavors and fragrances
food and beverages
glass & ceramic
personal care
pharmaceutical

format

mixture

Looking for similar products? Visit Product Comparison Guide

General description

This product is a certified reference material (CRM) certified to international standards BS EN ISO / IEC 17025 and ISO 17034 under UKAS accreditation.

Please note that the value presented on this page is nominal. All certified values for the current lots of all Paragon ASTM colour reference standards can be found on the Paragon Scientific webpage.

Application

Colour Reference Standard ASTM Colour 5 is ideal for regular checking of colour measuring equipment, validation of test results and inter-laboratory or inter-instrument comparison.
It′s used with colorimeters to analyze the colour control of products such as edible and industrial oils, fuels, chemicals, pharmaceuticals, paints and coatings.

Features and Benefits

  • Liquid colour standard compliant to ASTM D 6045 and D1500
  • Certified as per the dual UKAS accreditation to BS EN ISO / IEC 17025 and ISO 17034
  • Traceable to internationally recognized standards with high accuracy
  • Delivered in 500-mL tamper-evident bottles with safety data sheet
  • High-stability material with 18-months shelf life

Legal Information

ASTM is a registered trademark of American Society for Testing and Materials

pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Asp. Tox. 1

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service