12809
O-(Benzotriazol-1-yl)-N,N,N′,N′-bis(tetramethylene)uronium hexafluorophosphate
≥98.0%
Synonym(s):
(Benzotriazol-1-yloxy)dipyrrolidinocarbenium hexafluorophosphate, HBPyU
Select a Size
About This Item
Recommended Products
Quality Level
assay
≥98.0%
form
solid
reaction suitability
reaction type: Coupling Reactions
mp
176 °C (dec.) (lit.)
solubility
acetonitrile: 0.1 g/mL, clear
application(s)
peptide synthesis
storage temp.
2-8°C
SMILES string
F[P-](F)(F)(F)(F)F.C1CCN(C1)C(\On2nnc3ccccc23)=[N+]4/CCCC4
InChI
1S/C15H20N5O.F6P/c1-2-8-14-13(7-1)16-17-20(14)21-15(18-9-3-4-10-18)19-11-5-6-12-19;1-7(2,3,4,5)6/h1-2,7-8H,3-6,9-12H2;/q+1;-1
InChI key
XKTRAGMCMJYRRN-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Reagent for peptide coupling
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service