Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

12750

Sigma-Aldrich

Benzophenone

purum, ≥99.0% (GC)

Synonym(s):

Diphenyl ketone, Diphenylmethanone, NSC 8077

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5)2CO
CAS Number:
Molecular Weight:
182.22
Beilstein/REAXYS Number:
1238185
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

vapor pressure

1 mmHg ( 108 °C)

grade

purum

assay

≥99.0% (GC)

form

flakes

bp

305 °C (lit.)

mp

47-49 °C
47-51 °C (lit.)

SMILES string

O=C(C1=CC=CC=C1)C2=CC=CC=C2

InChI

1S/C13H10O/c14-13(11-7-3-1-4-8-11)12-9-5-2-6-10-12/h1-10H

InChI key

RWCCWEUUXYIKHB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Health hazard

signalword

Danger

Hazard Classifications

Aquatic Chronic 3 - Carc. 1B - STOT RE 2 Oral

target_organs

Liver,Kidney

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 1

flash_point_f

280.4 °F - closed cup

flash_point_c

138 °C - closed cup

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A powerful tool for graphene functionalization: Benzophenone mediated UV-grafting.
Roppolo I, et al.
Carbon, 77, 226-235 (2014)
Synthesis of main chain polymeric benzophenone photoinitiator via thiol-ene click chemistry and Its use in free radical polymerization.
Temel G, et al.
Journal of Polymer Science Part A: Polymer Chemistry, 48(23), 5306-5312 (2010)
Photopolymerization and photophysical properties of amine linked benzophenone photoinitiator for free radical polymerization.
Temel G, et al.
Journal of Photochemistry and Photobiology A: Chemistry, 219(1), 26-31 (2011)
Preparation and properties of poly (methacrylamide) s containing oligoaniline side chains.
Chen R and Benicewicz BC.
Macromolecules, 36(17), 6333-6339 (2003)
Feifan Yu et al.
PloS one, 8(2), e56597-e56597 (2013-02-21)
Affinity proteins binding to antibody constant regions have proved to be invaluable tools in biotechnology. Here, protein engineering was used to expand the repertoire of available immunoglobulin binding proteins via improvement of the binding strength between the widely used staphylococcal

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service