Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

12655

Supelco

Sieve Standard, traceable to NIST SRM

45 μm (325 mesh)

Synonym(s):

Sieve calibration standard, 45 μm (325 mesh)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

analytical standard

form

particles

manufacturer/tradename

Whitehouse Scientific Ltd SS395

particle size

45 μm (325 mesh)

application(s)

glass & ceramic
industrial qc
pharmaceutical

format

neat

Looking for similar products? Visit Product Comparison Guide

General description

Sieve Standard, traceable to NIST SRM is designed to find the particle size distribution (PSD) of a particle system.
The particle diameter of 45 μm (325 mesh) is measured using electroformed sieves and glass microsphere method.

Application

Used to standardize particle size analyzers and particle counters.
It′s suitable for use as a sieve calibration standard in electroformed sieve particle size analysis.[1]

Features and Benefits

  • suitable for routine instrument calibration checks, testing and corrections
  • traceable to NIST and SRM standards
  • available as a neat sample

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sieve Calibration-a New Simple but High Precision Approach
Rideal GR, et al.
Particle & Particle Systems Characterization, 17(2), 77-82 (2000)

Related Content

Particle size conversion table from our Lab Basics Technical Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service