Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

12183

Sigma-Aldrich

Disodium hydrogenphosphate 0.375M - Dipotassium-hydrogenphosphate 0.1875M - Tris pH 8, 100mM solution

Synonym(s):

Crystallization solution 48/Kit-No 02528

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

pH

8.0±0.2 ( in neat)

Quality Level

density

1.08 g/mL at 20 °C

storage temp.

2-8°C

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Giacomo Mancini et al.
Cell reports, 26(11), 2849-2858 (2019-03-14)
We found that exposure of adult animals to calorie-dense foods rapidly abolished expression of mitofusin 2 (Mfn2), a gene promoting mitochondrial fusion and mitochondrion-endoplasmic reticulum interactions, in white and brown fat. Mfn2 mRN was also robustly lower in obese human

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service