Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

11844

Sigma-Aldrich

Barium sulfate

tested according to Ph. Eur.

Synonym(s):

Barii sulfas, Baryte

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BaSO4
CAS Number:
Molecular Weight:
233.39
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
solubility:
water: insoluble

agency

USP/NF
tested according to Ph. Eur.

solubility

water: insoluble

SMILES string

[Ba++].[O-]S([O-])(=O)=O

InChI

1S/Ba.H2O4S/c;1-5(2,3)4/h;(H2,1,2,3,4)/q+2;/p-2

InChI key

TZCXTZWJZNENPQ-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Storage Class

13 - Non Combustible Solids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Camilleri et al.
International endodontic journal, 43(1), 21-30 (2009-11-07)
To identify the suitable ratio of alternative radiopacifiers to impart the necessary radiopacity to calcium silicate cements (CSC) and assess the purity of the radiopacifying agents. Alternative radiopacifying materials for incorporation into CSC included barium sulphate, titanium oxide, zinc oxide
LOCATION AND MECHANISM OF AIRWAY CONSTRICTION AFTER BARIUM SULFATE MICROEMBOLISM.
J A NADEL et al.
Journal of applied physiology, 19, 387-394 (1964-05-01)
B P Barnett et al.
Molecular pharmaceutics, 3(5), 531-538 (2006-10-03)
Alginate-poly-L-lysine-alginate (APA) microcapsules have been explored as vehicles for therapeutic drug and cell delivery. The permselectivity of these capsules provides a unique means of controlled drug release and immunoisolation of encapsulated cells. Immunoisolation is especially attractive as it abrogates the
Saeid Shafieiyoun et al.
Environmental science and pollution research international, 27(11), 11842-11854 (2020-01-26)
Engineered sulfate application has been proposed as an effective remedy to enhance the rate-limited biodegradation of petroleum-hydrocarbon-contaminated subsurface environments, but the effects of dissolved organic phase composition and salinity on the efficiency of this method are unknown. A series of
Joel A Feekes et al.
Brain : a journal of neurology, 129(Pt 8), 2189-2201 (2006-07-04)
The basal ganglia (BG) contain several functional compartments and multiple, parallel segregated circuits processing different cortical information through cortical-BG-thalamus-cortical loops. Three zones of corticostriatal input are present: sensorimotor, associative and limbic, which correspond to poor, intermediate and strong calbindin (CB)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service