11539
Azide Glucose Broth
suitable for microbiology
Synonym(s):
Azide Dextrose Broth
About This Item
Recommended Products
sterility
non-sterile
Quality Level
form
solid
shelf life
limited shelf life, expiry date on the label
composition
casein peptone, 15 g/L
D(+)-glucose, 7.5 g/L
meat extract, 4.8 g/L
sodium azide, 0.2 g/L
sodium chloride, 7.5 g/L
final pH
7.2±0.2 (25 °C)
application(s)
clinical testing
environmental
food and beverages
microbiology
suitability
selective for Enterococcus spp.
selective for Streptococcus spp.
Application
Other Notes
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service