Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

11355

Supelco

Aldicarb-(N-methyl-13C,d3, carbamoyl-13C) sulfone

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13C2C5D3H11N2O4S
CAS Number:
Molecular Weight:
227.27
MDL number:
UNSPSC Code:
77101502
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

assay

≥98.5% (HPLC)

shelf life

limited shelf life, expiry date on the label

mp

132-135 °C (lit.)

application(s)

agriculture
environmental

format

neat

mass shift

M+5

storage temp.

−20°C
−20°C

SMILES string

[2H][13C]([2H])([2H])N[13C](=O)O\N=C\C(C)(C)S(C)(=O)=O

InChI

1S/C7H14N2O4S/c1-7(2,14(4,11)12)5-9-13-6(10)8-3/h5H,1-4H3,(H,8,10)/b9-5+/i3+1D3,6+1

InChI key

YRRKLBAKDXSTNC-FONWFMSDSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Aldicarb-(N-methyl-13C,d3, carbamoyl-13C) sulfone is an isotope-labeled analog of aldicarb, a carbamate pesticide, used as an insecticide and nematicide[1] for the control of pests in agriculture.[2]

application

Isotope-labeled compounds are increasingly used in isotope dilution mass spectrometry (IDMS) for the quantitative analysis of pesticides. The main advantage of using the IDMS technique is because isotope-labeled compounds have nearly the same physical properties as their non-labeled counterpart analog, and thus show identical behavior during the workup and sample preparation process. This helps in overcoming the problems of matrix effects generally encountered in the usual LC-MS/GC-MS analysis potentially resulting in biased results. By spiking the sample before workup with its isotope labeled analog, the loss of analyte that leads to matrix effects can be determined and compensated.

Analysis Note

Isotopic enrichment atom%: ≥ 98.0

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

pictograms

Skull and crossbones

signalword

Danger

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chronic exposure to aldicarb-contaminated groundwater and human immune function
Fiore CM, et al.
Environmental Research, 41(2), 633-645 (1986)
Aldicarb poisoning
Ragouc-Sengler C, et al.
Human & Experimental Toxicology, 19(12), 657-662 (2000)
Determination of aldicarb, aldicarb oxime, and aldicarb nitrile in water by gas chromatography/mass spectrometry.
Trehy ML, et al.
Analytical Chemistry, 56(8), 1281-1285 (1984)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service