11309
5-(Acetylamino)-2-hydroxybenzoic acid
analytical standard
Synonym(s):
5-Acetamidosalicylic acid, N-Acetyl-5-aminosalicylic acid, Acetylmesalazine, NSC 54183
About This Item
Recommended Products
grade
analytical standard
Quality Level
assay
≥98.0% (GC)
shelf life
limited shelf life, expiry date on the label
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
application(s)
pharmaceutical
format
neat
storage temp.
2-8°C
SMILES string
CC(=O)Nc1ccc(O)c(c1)C(O)=O
InChI
1S/C9H9NO4/c1-5(11)10-6-2-3-8(12)7(4-6)9(13)14/h2-4,12H,1H3,(H,10,11)(H,13,14)
InChI key
GEFDRROBUCULOD-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Recommended products
signalword
Warning
hcodes
pcodes
Hazard Classifications
Acute Tox. 4 Oral - Eye Irrit. 2
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service