Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

10882

Sigma-Aldrich

N,N-Dimethyl-2-chloroacetoacetamide

Wacker Chemie AG, ≥95% (GC)

Synonym(s):

DMCAA, 2-Chloro-N,N-dimethylacetoacetamide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COCHClCON(CH3)2
CAS Number:
Molecular Weight:
163.60
Beilstein/REAXYS Number:
1761325
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

assay

≥95% (GC)

manufacturer/tradename

Wacker Chemie AG

refractive index

n20/D 1.484 (lit.)
n/D (lit.)

bp

95 °C/0.7 mmHg (lit.)

density

1.203 g/mL at 25 °C (lit.)

SMILES string

CN(C)C(=O)C(Cl)C(C)=O

InChI

1S/C6H10ClNO2/c1-4(9)5(7)6(10)8(2)3/h5H,1-3H3

InChI key

QJACRCAPQIYMIH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

N,N-Dimethyl-2-chloroacetoacetamide may be used in the synthesis of bidrin insecticide[1].

Other Notes

prices for bulk quantities on request

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

158.0 °F

flash_point_c

70 °C

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of 3-hydroxy-N, N-dimethyl-cis-crotonamide dimethyl phosphate and its N-methyl analog labeled with 32P and 14C.
Burton WB.
Journal of Labelled Compounds, 7(2), 111-125 (1971)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service