Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

10370

Sigma-Aldrich

trans-Anethole

purum, ≥98.0% (GC)

Synonym(s):

4-Propenylanisole, trans-1-Methoxy-4-(1-propenyl)benzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH=CHC6H4OCH3
CAS Number:
Molecular Weight:
148.20
Beilstein/REAXYS Number:
774229
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

purum

assay

≥98.0% (GC)

form

chips

refractive index

n20/D 1.561 (lit.)
n20/D 1.561

bp

234-237 °C (lit.)

mp

20-21 °C (lit.)

solubility

soluble, colorless to very faintly yellow

density

0.988 g/mL at 25 °C (lit.)

SMILES string

COc1ccc(\C=C\C)cc1

InChI

1S/C10H12O/c1-3-4-9-5-7-10(11-2)8-6-9/h3-8H,1-2H3/b4-3+

InChI key

RUVINXPYWBROJD-ONEGZZNKSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Naturally occurring phenylpropene derivative that is estrogenic at lower concentrations and cytotoxic at higher concentrations to cancer cell lines. The cytotoxicity is related to the metabolism of trans-anethole to 4-hydroxy-1-propenylbenzene.

pictograms

Exclamation markEnvironment

signalword

Warning

hcodes

Hazard Classifications

Aquatic Chronic 2 - Skin Sens. 1

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

213.8 °F

flash_point_c

101 °C

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hong-Ik Cho et al.
Journal of natural products, 76(9), 1717-1723 (2013-08-22)
The aim of this study was to investigate the hepatoprotective effect of anethole (trans-anethole, 1), a major component of Foeniculum vulgare, and its molecular mechanism during ischemia/reperfusion (I/R). Mice were subjected to 60 min of partial hepatic ischemia followed by
Josenilda Malveira Cavalcanti et al.
Journal of ethnopharmacology, 144(2), 240-247 (2012-09-25)
Croton zehntneri is a Euphorbiaceae species native to northeastern Brazil, where teas and beverages made from Croton zehntneri leaves are used as healing agents. To our knowledge, there is no experimental study supporting this claim of pharmacological activity. Full-thickness excisional
Meher U Nessa et al.
Anticancer research, 32(11), 4843-4850 (2012-11-17)
Chemopreventative phytochemicals having antitumour and antioxidant properties can overcome problems of chemoresistance and nonspecific toxicity towards normal cells that are associated with platinum-based chemotherapy against cancer. These agents exert their effects by bringing into play numerous cellular proteins that in
Masahiro Yutani et al.
Phytotherapy research : PTR, 25(11), 1707-1713 (2011-07-02)
trans-Anethole (anethole), a major component of anise oil, has a broad antimicrobial spectrum with antimicrobial activity relatively weaker than those of well-known antibiotics, and significantly enhances the antifungal activity of polygodial and dodecanol against the baker's yeast Saccharomyces cerevisiae and
Talita Perdigão Domiciano et al.
Naunyn-Schmiedeberg's archives of pharmacology, 386(4), 331-338 (2012-12-20)
Anethole [1-methoxy-4-(1-propenyl)benzene] occurs naturally as a major component of the essential oil of star anise (Illicium verum Hook.f., family Illiciaceae), comprising more than 90 % of its volatile components. Studies showed that this substance has antioxidant, antibacterial, antifungal, and anesthetic properties.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service