Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

09865

Sigma-Aldrich

Fmoc-Lys(Fmoc)-Lys[Fmoc-Lys(Fmoc)]-Lys{Fmoc-Lys(Fmoc)-Lys[Fmoc-Lys(Fmoc)]}-β-Ala-O-Wang resin

extent of labeling: ~0.8 mmol/g total N loading

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.26

extent of labeling

~0.8 mmol/g total N loading

Quality Level

matrix

crosslinked with 1% DVB

particle size

200-400 mesh

application(s)

peptide synthesis

storage temp.

2-8°C

Other Notes

Resin-bound octa-branched core for synthesizing Multiple Antigenic peptides, MAP[1][2]

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J P Tam
Proceedings of the National Academy of Sciences of the United States of America, 85(15), 5409-5413 (1988-08-01)
A convenient and versatile approach to the direct synthesis of a peptide-antigen matrix by the solid-phase method is described. The approach is called the multiple antigen peptide system (MAP) and it utilizes a simple scaffolding of a low number of
D N Posnett et al.
The Journal of biological chemistry, 263(4), 1719-1725 (1988-02-05)
Peptide antigens used to generate site-specific antibodies to proteins are of interest in the development of vaccines. The need to conjugate them to a carrier protein for optimal immunogenicity results in a number of problems including a possible immune response

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service