Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

09803

Sigma-Aldrich

Boc-β-(2-furyl)-Ala-OH (dicyclohexylammonium) salt

purum, ≥98.0% (HPLC)

Synonym(s):

(S)-2-(Boc-amino)-3-(2-furyl)propionic acid (dicyclohexylammonium) salt, Boc-3-(2-furyl)-L-alanine (dicyclohexylammonium) salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H17NO5 · C12H23N
CAS Number:
Molecular Weight:
436.58
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

grade

purum

Quality Level

assay

≥98.0% (HPLC)

form

powder

reaction suitability

reaction type: Boc solid-phase peptide synthesis

color

light yellow

application(s)

peptide synthesis

SMILES string

C1CCC(CC1)NC2CCCCC2.CC(C)(C)OC(=O)N[C@@H](Cc3ccco3)C(O)=O

InChI

1S/C12H17NO5.C12H23N/c1-12(2,3)18-11(16)13-9(10(14)15)7-8-5-4-6-17-8;1-3-7-11(8-4-1)13-12-9-5-2-6-10-12/h4-6,9H,7H2,1-3H3,(H,13,16)(H,14,15);11-13H,1-10H2/t9-;/m0./s1

InChI key

QYEOQPFAYJEDFE-FVGYRXGTSA-N

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service